Please login to your online account to display your discounted pricing

(S)-3-Hydroxy-3-phenylpropanoic acid, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  412120010

 View more versions of this product

Catalog No. AC412120010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Needle-Like Crystals
Formula Weight 166.18
Quantity 1g
Color White to almost white
Specific Rotation Condition −22.50° (20°C c=4,CH3OH)
Melting Point 116° to 119°C
CAS Max % 100.0
Molecular Formula C9H10O3
Assay Percent Range 99%
Chemical Name or Material (S)-3-Hydroxy-3-phenylpropanoic acid
MDL Number MFCD00145262
CAS 36567-72-3
CAS Min % 98.5
Packaging Glass bottle
Specific Rotation -21.5° to -23.5° (20°C, 589nm) (c=4, CH3OH)
Infrared Spectrum Authentic

Specifications

Chemical Name or Material (S)-3-Hydroxy-3-phenylpropanoic acid
CAS 36567-72-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H10O3
Formula Weight 166.18
MDL Number MFCD00145262