Learn More
(S)-3-Fluoropyrrolidine hydrochloride, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 442740050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
(S)-3-Fluoropyrrolidine hydrochloride | |
96.0 | |
Authentic | |
C4H8FN·ClH | |
6.15 | |
LENYOXXELREKGZ-WCCKRBBISA-N | |
(3S)-3-fluoropyrrolidine;hydrochloride | |
16217739 | |
97% |
136725-53-6 | |
100.0 | |
96.0 to max. 104% min. (Argentometry) | |
5g | |
s-3-fluoropyrrolidine hydrochloride, s-+-3-fluoropyrrolidine hydrochloride, 3s-3-fluoropyrrolidine hydrochloride, 3s-+-3-fluoropyrrolidine hydrochloride, s-3-fluoro-pyrrolidine hcl, s ;-3-fluoropyrrolidine hydrochloride, 3s-3-fluoropyrrolidine, chloride, s-3-fluoropyrrolidine hcl | |
C1CNCC1F.Cl | |
125.57 | |
125.57 |
Chemical Identifiers
136725-53-6 | |
125.57 | |
s-3-fluoropyrrolidine hydrochloride, s-+-3-fluoropyrrolidine hydrochloride, 3s-3-fluoropyrrolidine hydrochloride, 3s-+-3-fluoropyrrolidine hydrochloride, s-3-fluoro-pyrrolidine hcl, s ;-3-fluoropyrrolidine hydrochloride, 3s-3-fluoropyrrolidine, chloride, s-3-fluoropyrrolidine hcl | |
(3S)-3-fluoropyrrolidine;hydrochloride |
C4H8FN·ClH | |
LENYOXXELREKGZ-WCCKRBBISA-N | |
16217739 | |
C1CNCC1F.Cl |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Warning
RUO â Research Use Only