Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-3-Dimethylaminopyrrolidine dihydrochloride, 97%

$94.35 - $261.68

Chemical Identifiers

Chemical Name or Material (S)-(-)-3-Dimethylaminopyrrolidine dihydrochloride
CAS 144043-20-9
Molecular Formula C6H14N2·2HCl
Formula Weight 187.11
MDL Number MFCD08561117
Synonym (S)-(+)-N,N-Dimethyl-3-pyrrolidinamine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3066103

 
alfa aesar™
H3066103
1g Each for $261.68

AAH30661MD

 
alfa aesar™
H30661MD
250mg Each for $94.35
Description & Specifications

Specifications

Formula Weight 187.11
Chemical Name or Material (S)-(-)-3-Dimethylaminopyrrolidine dihydrochloride
Assay 97%
Molecular Formula C6H14N2·2HCl
MDL Number MFCD08561117
CAS 144043-20-9
Synonym (S)-(+)-N,N-Dimethyl-3-pyrrolidinamine dihydrochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 176° to 181°C
Quantity 1g
Optical Rotation −3.7° (c=1 in Water)
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.