Please login to your online account to display your discounted pricing

(S)-(-)-3-(Dimethylamino)pyrrolidine, 97%, Acros Organics


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC426450050

 View more versions of this product

Catalog No. AC426450050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 114.19
Quantity 5g
Assay Percent Range 97%
Chemical Name or Material (S)-(-)-3-(Dimethylamino)pyrrolidine
Packaging Glass bottle
Molecular Formula C6H14N2
CAS 132883-44-4

Specifications

Chemical Name or Material (S)-(-)-3-(Dimethylamino)pyrrolidine
Assay Percent Range 97%
Molecular Formula C6H14N2
Formula Weight 114.19