missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-3-Chloro-1-phenyl-1-propanol, 98+%

Manufacturer:  Alfa Aesar™ H5484703

 View more versions of this product

Catalog No. AAH5484703



Specifications

Specifications

(S)-(-)-3-Chloro-1-phenyl-1-propanol
58°C to 60°C
100306-34-1
MFCD00013309
JZFUHAGLMZWKTF-VIFPVBQESA-N
(1S)-3-chloro-1-phenylpropan-1-ol
2777894
≥98%
−26° (c=1 in Chloroform)
1g
C9H11ClO
1s-3-chloro-1-phenyl-propan-1-ol
C1=CC=C(C=C1)C(CCCl)O
170.636
170.64

Chemical Identifiers

100306-34-1
170.636
JZFUHAGLMZWKTF-VIFPVBQESA-N
2777894
C1=CC=C(C=C1)C(CCCl)O
C9H11ClO
MFCD00013309
1s-3-chloro-1-phenyl-propan-1-ol
(1S)-3-chloro-1-phenylpropan-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents