missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-3-Chloro-1,2-propanediol Acetonide, ACROS Organics™

Chemical Identifiers

CAS 60456-22-6
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.602
MDL Number MFCD00273365
InChI Key BNPOTXLWPZOESZ-RXMQYKEDSA-N
Synonym s-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, s-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, 4s-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, s---4-chloromethyl-2,2-dimethyl-1,3-dioxolane, s-+-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, s---4-chloromethyl-2,2-dimethyl-1,3-dioxolane, 1,3-dioxolane, 4-chloromethyl-2,2-dimethyl-, s, s-3-chloro-1,2-propanediol acetonide, pubchem14076, bnpotxlwpzoesz-rxmqykedsa
PubChem CID 6930483
IUPAC Name (4S)-4-(chloromethyl)-2,2-dimethyl-1,3-dioxolane
SMILES CC1(OCC(O1)CCl)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC361270010 SDS View Documents Acros Organics
AC361270010
1g Glass Bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

60456-22-6
150.602
BNPOTXLWPZOESZ-RXMQYKEDSA-N
6930483
CC1(OCC(O1)CCl)C
C6H11ClO2
MFCD00273365
s-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, s-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, 4s-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, s---4-chloromethyl-2,2-dimethyl-1,3-dioxolane, s-+-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, s---4-chloromethyl-2,2-dimethyl-1,3-dioxolane, 1,3-dioxolane, 4-chloromethyl-2,2-dimethyl-, s, s-3-chloro-1,2-propanediol acetonide, pubchem14076, bnpotxlwpzoesz-rxmqykedsa
(4S)-4-(chloromethyl)-2,2-dimethyl-1,3-dioxolane

Specifications

1.0950g/mL
Glass Bottle
−42.00
1g
98%
MFCD00273365
BNPOTXLWPZOESZ-RXMQYKEDSA-N
(4S)-4-(chloromethyl)-2,2-dimethyl-1,3-dioxolane
6930483
98%
50°C
1.4330 to 1.4350
63.0°C (37.0 mmHg)
60456-22-6
C6H11ClO2
s-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, s-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, 4s-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, s---4-chloromethyl-2,2-dimethyl-1,3-dioxolane, s-+-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, s---4-chloromethyl-2,2-dimethyl-1,3-dioxolane, 1,3-dioxolane, 4-chloromethyl-2,2-dimethyl-, s, s-3-chloro-1,2-propanediol acetonide, pubchem14076, bnpotxlwpzoesz-rxmqykedsa
CC1(OCC(O1)CCl)C
150.602
150.61
(S)-(-)-3-Chloro-1, 2-propanediol acetonide
Safety and Handling

Safety and Handling

TSCA : TSCA