missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-3-Chloro-1,2-propanediol Acetonide, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC361270050

 View more versions of this product

Catalog No. AC361270050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

150.61
C6H11ClO2
(S)-(-)-4-(Chloromethyl)-2, 2-dimethyl-1, 3-dioxolane
Glass Bottle
60456-22-6
5g
(S)-(-)-3-Chloro-1, 2-propanediol acetonide
MFCD00273365
98%

Chemical Identifiers

98%
150.61
(S)-(-)-4-(Chloromethyl)-2, 2-dimethyl-1, 3-dioxolane
C6H11ClO2
MFCD00273365
(S)-(-)-3-Chloro-1, 2-propanediol acetonide

Safety and Handling

SDS & Safety Documents

TSCA : TSCA