Please login to your online account to display your discounted pricing

(S)-3-Carboxy-4-hydroxyphenylglycine, Tocris

Group I antagonist/group II agonist

$479.45 - $1,954.04

Chemical Identifiers

Chemical Name or Material (S)-3-Carboxy-4-hydroxyphenylglycine
Assay Percent Range >98%
Molecular Formula C9H9NO5
Formula Weight 211.17
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

032950

 
r & d systems
RDS032950
50mg Each for $1,954.04

032910

 
r & d systems
RDS032910
10mg Each for $479.45
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 211.17
Chemical Name or Material (S)-3-Carboxy-4-hydroxyphenylglycine
Assay Percent Range >98%
Molecular Formula C9H9NO5
CAS 55136-48-6
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature