Please login to your online account to display your discounted pricing

(S)-3-bromo-α-methylbenzyl alcohol, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (S)-3-Bromo-α-methylbenzyl alcohol
CAS 134615-22-8
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C8H9BrO
Linear Formula CH3CH(OH)C6H4Br
Formula Weight 201.06
MDL Number MFCD06201859
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC376502500

  SDS
thermo fisher scientific
AC376502500
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC376500010

  SDS
fisher scientific acros
AC376500010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Liquid
Formula Weight 201.06
Quantity 250mg
Color Clear colorless
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material (S)-3-Bromo-α-methylbenzyl alcohol
Molecular Formula C8H9BrO
Linear Formula CH3CH(OH)C6H4Br
MDL Number MFCD06201859
Enantiomeric Excess 97.5% min. (GC)
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
CAS 134615-22-8
Synonym (S)-1-(3-Bromophenyl)ethanol
Infrared Spectrum Authentic
Assay Percent Range 95%
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.