Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-3-(Boc-amino)pyrrolidine, 99%, ee 99%

Chemical Identifiers

Chemical Name or Material (S)-(-)-3-(Boc-amino)pyrrolidine
CAS 122536-76-9
Molecular Formula C9H18N2O2
Formula Weight 186.25
MDL Number MFCD00143194
UN Number UN3259
Beilstein 5377813
Synonym (S)-3-(tert-Butoxycarbonylamino)pyrrolidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1969303

  SDS
alfa aesar™
L1969303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1969306

  SDS
alfa aesar™
L1969306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 186.25
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Chemical Name or Material (S)-(-)-3-(Boc-amino)pyrrolidine
MDL Number MFCD00143194
CAS 122536-76-9
Beilstein 5377813
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
Assay 99%
Optical Rotation −23° (c=1 in Methanol)
Health Hazard 1 Danger
Recommended Storage 2° to 8°C
Synonym (S)-3-(tert-Butoxycarbonylamino)pyrrolidine
Assay Percent Notes ee 99%
Quantity 1g
Melting Point 50° to 54°C
Molecular Formula C9H18N2O2
UN Number UN3259
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

2° to 8°C