Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-3-(Boc-amino)piperidine, 97%

$115.45 - $115.45

Chemical Identifiers

Chemical Name or Material (S)-3-(Boc-amino)piperidine
CAS 216854-23-8
Molecular Formula C10H20N2O2
Formula Weight 200.28
MDL Number MFCD03093383
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2695703

  SDS
alfa aesar™
H2695703
1g Each for $115.45
Description & Specifications

Specifications

Formula Weight 200.28
Chemical Name or Material (S)-3-(Boc-amino)piperidine
Assay 97%
Molecular Formula C10H20N2O2
MDL Number MFCD03093383
CAS 216854-23-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 122° to 127°C
Quantity 1g
Optical Rotation −15.0° (c=1 in MeOH)

Applications
(S)-3-(Boc-amino)piperidine, is used as an intermediate for pigments. It is also used as important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also an useful intermediate for the synthesis of a variety of chiral aminopiperidinyl quinolones as potent antibacterial agents against resistant pathogens; and alkynylpyrimidine amide derivatives as orally available inhibitors of Tie-2 kinase.

Solubility
Freely soluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.