Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde, 95%

$90.81 - $223.99

Chemical Identifiers

Chemical Name or Material (S)-(-)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde
CAS 102308-32-7
Molecular Formula C11H19NO4
Formula Weight 229.27g/mol
MDL Number MFCD00209557
Beilstein 3591324
Merck Index 14,4371
Synonym (S)-(-)-3-tert-Butoxycarbonyl-2,2-dimethyloxazolidine-4-carboxaldehyde; Garner's Aldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5104903

 
alfa aesar™
H51049-03
1g Each for $223.99

AAH51049MD

 
alfa aesar™
H51049-MD
250mg Each for $90.81
Description & Specifications

Specifications

Quantity 1g
Quantity 1g
Optical Rotation −90° (c=1 in Chloroform)
Chemical Name or Material (S)-(-)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde
CAS 102308-32-7
Assay 95%
Molecular Formula C11H19NO4
Formula Weight 229.27g/mol
MDL Number MFCD00209557
Beilstein 3591324
Merck Index 14,4371
Synonym (S)-(-)-3-tert-Butoxycarbonyl-2,2-dimethyloxazolidine-4-carboxaldehyde; Garner's Aldehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C

Applications
It is used to prepare γ-aminophosphonates (N-arylamide phosphonates as subtype-selective sphingosine 1-phosphate receptor agonists and antagonists. It is also used to synthesize N-sulfonylated propargylglycine derivatives as metalloproteinase inhibitors. Also widely utilized in the asymmetric synthesis of natural products.

Solubility
Slightly soluble in water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C