missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-3-Benzyloxycarbonylamino-2-(Boc-amino)propionic acid dicyclohexylammonium salt, 98%

Manufacturer:  Alfa Aesar™ H6155803

 View more versions of this product

Catalog No. AAH6155803



Specifications

Specifications

(S)-3-Benzyloxycarbonylamino-2-(Boc-amino)propionic acid dicyclohexylammonium salt
65710-58-9
MFCD00236880
a-boc-n
CC(C)(C)OC(=O)NC(CNC(=O)OCC1=CC=CC=C1)C(=O)O.C1CCC(CC1)NC2CCCCC2
519.683
385.54
98%
1g
C28H45N3O6
4100906
RPWGTQRQPVPFKR-YDALLXLXSA-N
N-cyclohexylcyclohexanamine;(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(phenylmethoxycarbonylamino)propanoic acid
2755949
Solid

Chemical Identifiers

65710-58-9
519.683
RPWGTQRQPVPFKR-YDALLXLXSA-N
2755949
CC(C)(C)OC(=O)NC(CNC(=O)OCC1=CC=CC=C1)C(=O)O.C1CCC(CC1)NC2CCCCC2
C28H45N3O6
MFCD00236880
a-boc-n
N-cyclohexylcyclohexanamine;(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(phenylmethoxycarbonylamino)propanoic acid
Documents