Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-3-(Benzyloxycarbonyl)oxazolidine-4-carboxylic acid, 98%

$76.82 - $233.97

Chemical Identifiers

Chemical Name or Material (S)-(-)-3-(Benzyloxycarbonyl)oxazolidine-4-carboxylic acid
CAS 97534-82-2
Molecular Formula C12H13NO5
Formula Weight 251.24
MDL Number MFCD00274193
Synonym N-Cbz-(S)-oxaproline; Z-OxaPro
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2705603

 
alfa aesar™
H2705603
1g Each for $76.82

AAH2705606

 
alfa aesar™
H2705606
5g Each for $233.97
Description & Specifications

Specifications

Formula Weight 251.24
Chemical Name or Material (S)-(-)-3-(Benzyloxycarbonyl)oxazolidine-4-carboxylic acid
Assay 98%
Molecular Formula C12H13NO5
MDL Number MFCD00274193
CAS 97534-82-2
Synonym N-Cbz-(S)-oxaproline; Z-OxaPro
Melting Point 78° to 81°C
Quantity 1g
Optical Rotation −79° (c=1 in Chloroform)