Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-3-Aminopyrrolidine dihydrochloride, 98%

$49.72 - $138.39

Chemical Identifiers

Chemical Name or Material (S)-(+)-3-Aminopyrrolidine dihydrochloride
CAS 116183-83-6
Molecular Formula C4H10N2·2HCl
Formula Weight 159.06
MDL Number MFCD00070605
Beilstein 4870016
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1222203

 
alfa aesar™
L1222203
1g Each for $138.39

AAL12222MD

 
alfa aesar™
L12222MD
250mg Each for $49.72
Description & Specifications

Specifications

Formula Weight 159.06
Chemical Name or Material (S)-(+)-3-Aminopyrrolidine dihydrochloride
Assay 98%
Molecular Formula C4H10N2·2HCl
MDL Number MFCD00070605
CAS 116183-83-6
Beilstein 4870016
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Optical Rotation +1.2° (c=10 in Water)
Health Hazard 1 Warning
Melting Point ≈283°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.