missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-3-Aminopyrrolidine dihydrochloride, 98%

Manufacturer:  Alfa Aesar™ L12222MD

 View more versions of this product

Catalog No. AAL12222MD



Specifications

Specifications

(S)-(+)-3-Aminopyrrolidine dihydrochloride
~283°C
116183-83-6
MFCD00070605
s-+-3-aminopyrrolidine dihydrochloride, s-pyrrolidin-3-amine dihydrochloride, 3s-+-3-aminopyrrolidine dihydrochloride, 3s-pyrrolidin-3-amine dihydrochloride, s-3-aminopyrrolidine 2hcl, s-3-aminopyrrolidine dihydrochloride, s-+-3-aminopyrrolidine 2hcl, 3-pyrrolidinamine, dihydrochloride, 3s, pubchem11156, s-+-3-aminopyrrolidinedihydrochloride
C1CNCC1N.Cl.Cl
159.054
159.06
+1.2° (c=10 in Water)
250mg
C4H12Cl2N2
4870016
NJPNCMOUEXEGBL-FHNDMYTFSA-N
(3S)-pyrrolidin-3-amine;dihydrochloride
13500644
98%

Chemical Identifiers

116183-83-6
159.054
NJPNCMOUEXEGBL-FHNDMYTFSA-N
13500644
C1CNCC1N.Cl.Cl
C4H12Cl2N2
MFCD00070605
s-+-3-aminopyrrolidine dihydrochloride, s-pyrrolidin-3-amine dihydrochloride, 3s-+-3-aminopyrrolidine dihydrochloride, 3s-pyrrolidin-3-amine dihydrochloride, s-3-aminopyrrolidine 2hcl, s-3-aminopyrrolidine dihydrochloride, s-+-3-aminopyrrolidine 2hcl, 3-pyrrolidinamine, dihydrochloride, 3s, pubchem11156, s-+-3-aminopyrrolidinedihydrochloride
(3S)-pyrrolidin-3-amine;dihydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents