Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-3-Aminopyrrolidine, 99%, ee 99%

Chemical Identifiers

Chemical Name or Material (S)-(-)-3-Aminopyrrolidine
CAS 128345-57-3
Molecular Formula C4H10N2
Formula Weight 86.14
MDL Number MFCD00143193
UN Number UN2735
Beilstein 4290648
Synonym (S)-(-)-3-Pyrrolidinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1950003

  SDS
alfa aesar™
L1950003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1950006

  SDS
alfa aesar™
L1950006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 86.14
Chemical Name or Material (S)-(-)-3-Aminopyrrolidine
Assay 99%
Assay Percent Notes ee 99%
Molecular Formula C4H10N2
MDL Number MFCD00143193
CAS 128345-57-3
UN Number UN2735
Synonym (S)-(-)-3-Pyrrolidinamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Boiling Point 159° to 160°C
Beilstein 4290648
Quantity 1g
Flash Point 68°C (154°F)
Optical Rotation −20° (Neat)
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.