Please login to your online account to display your discounted pricing

(S)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride, 95%, 98% ee, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  362040010

 View more versions of this product

Catalog No. AC362040010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94% (HPLC)
Formula Weight 229.71
Physical Form Powder
Quantity 1g
Color White
Chemical Name or Material (S)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride
Molecular Formula C11H15NO2·HCl
Health Hazard 1 Warning
Enantiomeric Excess 97.5% (GC) min.
Assay Percent Range 95%
Infrared Spectrum Authentic
CAS 167834-24-4

Specifications

Chemical Name or Material (S)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride
Assay ≥94% (HPLC)
Assay Percent Range 95%
Molecular Formula C11H15NO2·HCl
Formula Weight 229.71
Accessories

Warning

SDS & Safety Documents