Please login to your online account to display your discounted pricing

(S)-3-Amino-3-phenylpropan-1-ol, 95%, 98% e.e., ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  362172500

 View more versions of this product

Catalog No. AC362172500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Viscous Liquid or Low Melting Solid
Formula Weight 151.21
Quantity 250mg
Color Colorless or white
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material (S)-3-Amino-3-phenylpropan-1-ol
Assay Percent Range 95%
MDL Number MFCD01311768
Synonym (S)-1-Phenyl-3-propanolamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 95%, (98% e.e.)
Molecular Formula C9H13NO
CAS 82769-76-4
Enantiomeric Excess 97.5% min.
Packaging Glass bottle
Infrared Spectrum Authentic

Specifications

Chemical Name or Material (S)-3-Amino-3-phenylpropan-1-ol
Name Note 95%, (98% e.e.)
CAS 82769-76-4
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C9H13NO
Formula Weight 151.21
MDL Number MFCD01311768
Synonym (S)-1-Phenyl-3-propanolamine
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents