missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-3-Amino-3-phenylpropan-1-ol, 95%, 98% ee, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 362170010

 View more versions of this product

Catalog No. AC362170010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

(S)-3-Amino-3-phenylpropan-1-ol
Authentic
Colorless or White
95%, (98% e.e.)
94.0
94% min. (GC)
MFCD01311768
SEQXIQNPMQTBGN-VIFPVBQESA-N
(3S)-3-amino-3-phenylpropan-1-ol
2734520
Viscous Liquid or Low Melting Solid
97.5% min.
Glass bottle
1g
82769-76-4
100.0
C9H13NO
3s-3-amino-3-phenyl-propan-1-ol, 3s-3-amino-3-phenylpropan-1-ol, ksc491q5n, pubchem13854, s-1-phenyl-3-propanolamine, s-3-amino-3-phenyl-1-propanol, s-3-amino-3-phenyl-propan-1-ol, s-3-amino-3-phenylpropan-1-ol, s-3-phenyl-beta-alaninol, s-beta-phenylalaninol
C1=CC=C(C=C1)C(CCO)N
151.21
151.21
95%,98% ee

Chemical Identifiers

82769-76-4
151.21
SEQXIQNPMQTBGN-VIFPVBQESA-N
2734520
C1=CC=C(C=C1)C(CCO)N
C9H13NO
MFCD01311768
3s-3-amino-3-phenyl-propan-1-ol, 3s-3-amino-3-phenylpropan-1-ol, ksc491q5n, pubchem13854, s-1-phenyl-3-propanolamine, s-3-amino-3-phenyl-1-propanol, s-3-amino-3-phenyl-propan-1-ol, s-3-amino-3-phenylpropan-1-ol, s-3-phenyl-beta-alaninol, s-beta-phenylalaninol
(3S)-3-amino-3-phenylpropan-1-ol
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA

SDS Documents