Learn More
(S)-3-Amino-1-benzylpiperidine, 97%, Thermo Scientific Chemicals
$103.41 - $398.93
Chemical Identifiers
CAS | 168466-85-1 |
---|---|
Molecular Formula | C12H18N2 |
Molecular Weight (g/mol) | 190.29 |
MDL Number | MFCD03093375 |
InChI Key | HARWNWOLWMTQCC-LBPRGKRZSA-N |
Synonym | s-1-benzylpiperidin-3-amine, s-3-amino-1-benzylpiperidine, 3s-1-benzylpiperidin-3-amine, s-1-benzyl-3-aminopiperidine, s-3-amino-1-benzyl-piperidine, 3-piperidinamine, 1-phenylmethyl-, 3s, s-n-benzyl-3-amino-piperidine, s-1-benzyl-piperidine-3-ylamine, s-3-amino-n-benzylpiperidine, pubchem11490 |
PubChem CID | 854131 |
IUPAC Name | (3S)-1-benzylpiperidin-3-amine |
SMILES | C1CC(CN(C1)CC2=CC=CC=C2)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5277203
|
Thermo Scientific Chemicals
H5277203 |
1 g |
Each for $103.41
|
|
|||||
AAH5277206
|
Thermo Scientific Chemicals
H5277206 |
5 g |
Each for $398.93
|
|
|||||
Description
(S)-3-Amino-1-benzylpiperidine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-3-Amino-1-benzylpiperidine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Freely soluble in ethanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
168466-85-1 | |
190.29 | |
HARWNWOLWMTQCC-LBPRGKRZSA-N | |
854131 | |
C1CC(CN(C1)CC2=CC=CC=C2)N |
C12H18N2 | |
MFCD03093375 | |
s-1-benzylpiperidin-3-amine, s-3-amino-1-benzylpiperidine, 3s-1-benzylpiperidin-3-amine, s-1-benzyl-3-aminopiperidine, s-3-amino-1-benzyl-piperidine, 3-piperidinamine, 1-phenylmethyl-, 3s, s-n-benzyl-3-amino-piperidine, s-1-benzyl-piperidine-3-ylamine, s-3-amino-n-benzylpiperidine, pubchem11490 | |
(3S)-1-benzylpiperidin-3-amine |
Specifications
168466-85-1 | |
MFCD03093375 | |
s-1-benzylpiperidin-3-amine, s-3-amino-1-benzylpiperidine, 3s-1-benzylpiperidin-3-amine, s-1-benzyl-3-aminopiperidine, s-3-amino-1-benzyl-piperidine, 3-piperidinamine, 1-phenylmethyl-, 3s, s-n-benzyl-3-amino-piperidine, s-1-benzyl-piperidine-3-ylamine, s-3-amino-n-benzylpiperidine, pubchem11490 | |
HARWNWOLWMTQCC-LBPRGKRZSA-N | |
(3S)-1-benzylpiperidin-3-amine | |
854131 | |
97% |
C12H18N2 | |
1 g | |
Freely soluble in ethanol. | |
C1CC(CN(C1)CC2=CC=CC=C2)N | |
190.29 | |
190.29 | |
(S)-3-Amino-1-benzylpiperidine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only