missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-3-Amino-1,2-propanediol, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC435280010

 View more versions of this product

Catalog No. AC435280010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(S)-3-Amino-1,2-propanediol
113°C
Glass bottle
− 30.00
Colorless to White
1g
97.5
98%
1,2-propanediol, 3-amino-, 2s, 2s-2,3-dihydroxypropylamine, 2s-3-amino-1,2-propanediol, 2s-3-aminopropane-1,2-diol, ksc496i7p, s---3-amino-1,2-propandiol, s---3-amino-1,2-propanediol, s-2,3-dihydroxypropylamine, s-3-amino-1,2-propanediol, s-3-aminopropane-1,2-diol
C(C(CO)O)N
91.11
CHEBI:1456
Liquid To Low Melting Solid
1.1750g/mL
Authentic
1.175
128.0°C to 130.0°C (5.0mmHg)
55.0°C to 57.0°C
61278-21-5
100.0
C3H9NO2
KQIGMPWTAHJUMN-VKHMYHEASA-N
(2S)-3-aminopropane-1,2-diol
440902
91.11
98%

Chemical Identifiers

61278-21-5
91.11
1,2-propanediol, 3-amino-, 2s, 2s-2,3-dihydroxypropylamine, 2s-3-amino-1,2-propanediol, 2s-3-aminopropane-1,2-diol, ksc496i7p, s---3-amino-1,2-propandiol, s---3-amino-1,2-propanediol, s-2,3-dihydroxypropylamine, s-3-amino-1,2-propanediol, s-3-aminopropane-1,2-diol
CHEBI:1456
C(C(CO)O)N
C3H9NO2
KQIGMPWTAHJUMN-VKHMYHEASA-N
440902
(2S)-3-aminopropane-1,2-diol
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

SDS Documents