Please login to your online account to display your discounted pricing

(S)-3-Amino-1,2-propanediol, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (S)-3-Amino-1, 2-propanediol
CAS 61278-21-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (Total base)
Assay Percent Range 98%
Molecular Formula C3H9NO2
Formula Weight 91.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC435280010

  SDS
thermo fisher scientific
AC435280010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC435280050

  SDS
thermo fisher scientific
AC435280050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid To Low Melting Solid
Assay ≥97.5% (Total base)
Formula Weight 91.11
Quantity 1g
Color Colorless to white
Density 1.1750g/mL
Melting Point 53° to 57°C
CAS Max % 100.0
Specific Rotation −27° to −31° (20°C, 589nm) (c=2, 1M HCl)
Chemical Name or Material (S)-3-Amino-1, 2-propanediol
Assay Percent Range 98%
Molecular Formula C3H9NO2
CAS 61278-21-5
Infrared Spectrum Authentic
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.