Please login to your online account to display your discounted pricing

(S)-3-Amino-1,2-propanediol, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC435280050

 View more versions of this product

Catalog No. AC435280050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid To Low Melting Solid
Assay ≥97.5% (Total base)
Formula Weight 91.11
Quantity 5g
Color Colorless to white
Melting Point 53° to 57°C
CAS Max % 100.0
Chemical Name or Material (S)-3-Amino-1, 2-propanediol
Specific Rotation −27° to −31° (20°C, 589nm) (c=2, 1M HCl)
CAS 61278-21-5
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C3H9NO2
Infrared Spectrum Authentic

Specifications

Chemical Name or Material (S)-3-Amino-1, 2-propanediol
CAS 61278-21-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (Total base)
Assay Percent Range 98%
Molecular Formula C3H9NO2
Formula Weight 91.11
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.

SDS & Safety Documents