missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6351603
Description
(S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications
(S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid | |
C22H22N2O6 | |
1 g | |
fmoc-dap alloc-oh, a-allyloxycarbonyl-l-2,3-diaminopropion, fmoc-dap aloc-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-allyloxy carbonyl amino propanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-prop-2-en-1-yloxy carbonyl amino propanoic acid, ambotzfaa1366, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-prop-2-en-1-yloxy carbonyl amino propanoic acid, fmoc-dap alloc-oh hplc, fmoc-3-allyloxycarbonylamino-l-ala-oh, a-fmoc-n-, n-, a-fmoc-n- | |
C=CCOC(=O)NCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 | |
410.426 | |
410.42 | |
Solid |
188970-92-5 | |
MFCD00273468 | |
9161065 | |
MPVGCCAXXFLGIU-IBGZPJMESA-N | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(prop-2-enoxycarbonylamino)propanoic acid | |
2756107 | |
95% |
Chemical Identifiers
188970-92-5 | |
410.426 | |
MPVGCCAXXFLGIU-IBGZPJMESA-N | |
2756107 | |
C=CCOC(=O)NCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 |
C22H22N2O6 | |
MFCD00273468 | |
fmoc-dap alloc-oh, a-allyloxycarbonyl-l-2,3-diaminopropion, fmoc-dap aloc-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-allyloxy carbonyl amino propanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-prop-2-en-1-yloxy carbonyl amino propanoic acid, ambotzfaa1366, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-prop-2-en-1-yloxy carbonyl amino propanoic acid, fmoc-dap alloc-oh hplc, fmoc-3-allyloxycarbonylamino-l-ala-oh, a-fmoc-n-, n-, a-fmoc-n- | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(prop-2-enoxycarbonylamino)propanoic acid |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C; Store under Argon
RUO – Research Use Only