Please login to your online account to display your discounted pricing

(S)-3,4-DCPG, Tocris

Potent, selective mGlu8a agonist

$257.85 - $1,073.10

Chemical Identifiers

Chemical Name or Material (S)-3, 4-DCPG
Assay Percent Range >98%
Molecular Formula C10H9NO6
Formula Weight 239.18
Synonym (S)-3, 4-Dicarboxyphenylglycine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

130250

 
r & d systems
RDS130250
50mg Each for $1,073.10

130210

 
r & d systems
RDS130210
10mg Each for $257.85
Description & Specifications

Specifications

Formula Weight 239.18
Quantity 50mg
Chemical Name or Material (S)-3, 4-DCPG
CAS 201730-11-2
Assay Percent Range >98%
Molecular Formula C10H9NO6
Synonym (S)-3, 4-Dicarboxyphenylglycine
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature