Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-3,3'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl, 97%

$266.36 - $266.36

Chemical Identifiers

Chemical Name or Material (S)-3,3'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl
CAS 142010-87-5
Molecular Formula C24H20Br2O4
Formula Weight 532.23g/mol
MDL Number MFCD16294495
Synonym (S)-3,3'-Dibromo-1,1'-Bi(2-naphthol) bis(methoxymethyl)ether
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3178303

 
alfa aesar™
H31783-03
1g Each for $266.36
Description & Specifications

Specifications

Melting Point 93° to 95°C
Quantity 1g
Chemical Name or Material (S)-3,3'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl
CAS 142010-87-5
Assay 97%
Molecular Formula C24H20Br2O4
Formula Weight 532.23g/mol
MDL Number MFCD16294495
Synonym (S)-3,3'-Dibromo-1,1'-Bi(2-naphthol) bis(methoxymethyl)ether