Please login to your online account to display your discounted pricing

(S)-(-)-3,3'-Dibromo-1,1'-bi-2-naphthol 97+%, ACROS Organics™


Quantity 100mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC384661000

 View more versions of this product

Catalog No. AC384661000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.0%
Formula Weight 444.12
Physical Form Powder
Quantity 100mg
Color White to pale yellow-beige
Specific Rotation Condition −64.00 (20.00°C c=0,2%,chloroform)
CAS Max % 100.0
CAS Min % 97.0
Infrared Spectrum Authentic
Chemical Name or Material (S)-(-)-3, 3'-Dibromo-1, 1'-bi-2-naphthol
Assay Percent Range 97+%
Molecular Formula C20H12Br2O2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD03093629
CAS 119707-74-3
Specific Rotation -60° to -68° (20°C, 589nm) (c=1, CHCl3)
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material (S)-(-)-3, 3'-Dibromo-1, 1'-bi-2-naphthol
CAS 119707-74-3
CAS Min % 97.0
CAS Max % 100.0
Assay ≥97.0%
Assay Percent Range 97+%
Molecular Formula C20H12Br2O2
Formula Weight 444.12
MDL Number MFCD03093629
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents