Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-3,3'-Dibromo-1,1'-bi-2-naphthol, 96%

Chemical Identifiers

Chemical Name or Material (S)-(-)-3,3'-Dibromo-1,1'-bi-2-naphthol
CAS 119707-74-3
Formula Weight 444.12
MDL Number MFCD03093629
UN Number UN3077
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5544503

 
alfa aesar™
H5544503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55445MD

 
alfa aesar™
H55445MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 256° to 260°C
Formula Weight 444.12
Chemical Name or Material (S)-(-)-3,3'-Dibromo-1,1'-bi-2-naphthol
Physical Form Powder
Assay 96%
MDL Number MFCD03093629
CAS 119707-74-3
UN Number UN3077
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color White to brown
Quantity 1g
DOT Information Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.