Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol, 99%

$198.57 - $532.59

Chemical Identifiers

Chemical Name or Material (S)-(-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol
CAS 205927-03-3
Molecular Formula C24H34O2
Formula Weight 354.54
MDL Number MFCD01862439
Synonym (S)-BIPHEN-H2; (S)-3,3'-Di-tert-butyl-2,2'-dihydroxy-5,5',6,6'-tetramethylbiphenyl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2734203

 
alfa aesar™
H2734203
1g Each for $532.59

AAH27342MD

 
alfa aesar™
H27342MD
250mg Each for $198.57
Description & Specifications

Specifications

Formula Weight 354.54
Chemical Name or Material (S)-(-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol
Assay 99%
Molecular Formula C24H34O2
MDL Number MFCD01862439
CAS 205927-03-3
Synonym (S)-BIPHEN-H2; (S)-3,3'-Di-tert-butyl-2,2'-dihydroxy-5,5',6,6'-tetramethylbiphenyl
Melting Point 171° to 173°C
Quantity 1g
Optical Rotation −84° (c=0.35 in THF)