Please login to your online account to display your discounted pricing

S 25585, Tocris™

Potent and selective NPY Y5 receptor antagonist

$257.85 - $1,073.10

Chemical Identifiers

Chemical Name or Material S 25585
Assay Percent Range >98%
Molecular Formula C22H23F3N4O6S
Formula Weight 528.5
Synonym 1-Benzoyl-2-[[trans-4-[[[[2-nitro-4-(trifluoromethyl)phenyl]sulfonyl]amino]methyl]cyclohexyl]carbonyl]hydrazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

343210

 
r & d systems
RDS343210
10mg Each for $257.85

343250

 
r & d systems
RDS343250
50mg Each for $1,073.10
Description & Specifications

Specifications

Formula Weight 528.5
Quantity 10mg
Chemical Name or Material S 25585
Assay Percent Range >98%
Molecular Formula C22H23F3N4O6S
CAS 263849-50-9
Synonym 1-Benzoyl-2-[[trans-4-[[[[2-nitro-4-(trifluoromethyl)phenyl]sulfonyl]amino]methyl]cyclohexyl]carbonyl]hydrazine
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature