Please login to your online account to display your discounted pricing

S 24795, Tocris™

Partial agonist at α7 nAChR

$163.07 - $678.85

Chemical Identifiers

Chemical Name or Material S 24795
Assay Percent Range >98%
Molecular Formula C14H13BrINO
Formula Weight 418.07
Synonym 2-[2-(4-Bromophenyl)-2-oxoethyl]-1-methylpyridinium iodide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

351850

 
r & d systems
RDS351850
50mg Each for $678.85

351810

 
r & d systems
RDS351810
10mg Each for $163.07
Description & Specifications

Specifications

Chemical Name or Material S 24795
CAS 304679-75-2
Recommended Storage Store at +4°C
Quantity 50mg
Assay Percent Range >98%
Molecular Formula C14H13BrINO
Formula Weight 418.07
Synonym 2-[2-(4-Bromophenyl)-2-oxoethyl]-1-methylpyridinium iodide
Safety and Handling

Store at +4°C