Please login to your online account to display your discounted pricing

(S)-(-)-2-(tert-Butoxycarbonylamino)-3-phenyl-1-propanol, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (S)-(-)-2-(tert-Butoxycarbonylamino)-3-phenyl-1-propanol
CAS 66605-57-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C14H21NO3
Formula Weight 251.32
MDL Number MFCD00076976
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC334240050

  SDS
fisher scientific acros
334240050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 251.32
Melting Point >94°C
Quantity 5g
Color White to off-white
CAS Max % 100.0
Infrared Spectrum Authentic
CAS 66605-57-0
Specific Rotation -22° to -28° (20°C, 589nm) (c=1, CHCl3)
Chemical Name or Material (S)-(-)-2-(tert-Butoxycarbonylamino)-3-phenyl-1-propanol
Molecular Formula C14H21NO3
Assay Percent Range 98%
MDL Number MFCD00076976
CAS Min % 97.5
Packaging Glass bottle