Please login to your online account to display your discounted pricing

(S)-(-)-2-(tert-Butoxycarbonylamino)-3-phenyl-1-propanol, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  334240050

 View more versions of this product

Catalog No. AC334240050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Melting Point >94°C
Formula Weight 251.32
Physical Form Powder
Quantity 5g
Color White to off-white
CAS Max % 100.0
Infrared Spectrum Authentic
Specific Rotation -22° to -28° (20°C, 589nm) (c=1, CHCl3)
CAS 66605-57-0
Chemical Name or Material (S)-(-)-2-(tert-Butoxycarbonylamino)-3-phenyl-1-propanol
Molecular Formula C14H21NO3
Assay Percent Range 98%
MDL Number MFCD00076976
CAS Min % 97.5
Packaging Glass bottle

Specifications

Chemical Name or Material (S)-(-)-2-(tert-Butoxycarbonylamino)-3-phenyl-1-propanol
CAS 66605-57-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C14H21NO3
Formula Weight 251.32
MDL Number MFCD00076976