Please login to your online account to display your discounted pricing

(S)-(+)-2-Phenylglycine methyl ester hydrochloride, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (S)-(+)-2-Phenylglycine methyl ester hydrochloride
Assay Percent Range 97%
Molecular Formula C9H11NO2·ClH
Formula Weight 201.65
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC442190100

 
thermo fisher scientific
AC442190100
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC442190500

 
thermo fisher scientific
AC442190500
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 201.65
Quantity 10g
Chemical Name or Material (S)-(+)-2-Phenylglycine methyl ester hydrochloride
Assay Percent Range 97%
Molecular Formula C9H11NO2·ClH
CAS 15028-39-4