missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-(+)-2-Phenylglycine methyl ester hydrochloride, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC442190500
Specifications
(S)-(+)-2-Phenylglycine methyl ester hydrochloride | |
200°C | |
15028-39-4 | |
C9H11NO2·ClH | |
DTHMTBUWTGVEFG-QRPNPIFTSA-N | |
methyl (2S)-2-amino-2-phenylacetate;hydrochloride | |
11217910 | |
97% |
Glass Bottle | |
50g | |
96% min. (HPLC) | |
aminophenylacetic acid methyl ester, d---alpha-phenylglycine methyl ester hydrochloride, d-phenylglycine methyl ester hydrochloride, h-phg-ome hcl, h-phg-ome hydrochloride, h-phg-ome.hcl, methyl 2s-2-amino-2-phenylacetate hydrochloride, s-+-2-phenylglycine methyl ester hydrochloride, s-2-phenylglycine methyl ester hydrochloride, s-methyl 2-amino-2-phenylacetate hydrochloride | |
COC(=O)C(C1=CC=CC=C1)N.Cl | |
201.65 | |
201.65 |
Chemical Identifiers
15028-39-4 | |
201.65 | |
aminophenylacetic acid methyl ester, d---alpha-phenylglycine methyl ester hydrochloride, d-phenylglycine methyl ester hydrochloride, h-phg-ome hcl, h-phg-ome hydrochloride, h-phg-ome.hcl, methyl 2s-2-amino-2-phenylacetate hydrochloride, s-+-2-phenylglycine methyl ester hydrochloride, s-2-phenylglycine methyl ester hydrochloride, s-methyl 2-amino-2-phenylacetate hydrochloride | |
methyl (2S)-2-amino-2-phenylacetate;hydrochloride |
C9H11NO2·ClH | |
DTHMTBUWTGVEFG-QRPNPIFTSA-N | |
11217910 | |
COC(=O)C(C1=CC=CC=C1)N.Cl |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning