Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-2-(Phenylcarbamoyloxy)propionic acid, 99%

$33.15 - $245.61

Chemical Identifiers

Chemical Name or Material (S)-(-)-2-(Phenylcarbamoyloxy)propionic acid
CAS 102936-05-0
Molecular Formula C10H11NO4
Formula Weight 209.2
MDL Number MFCD00063138
Synonym (S)-(-)-Carbamalactic acid; (S)-(-)-2-(Phenylaminocarbonyloxy)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2542503

  SDS
alfa aesar™
B2542503
1g Each for $33.15

AAB2542509

  SDS
alfa aesar™
B2542509
10g Each for $245.61
Description & Specifications

Specifications

Formula Weight 209.2
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00063138
Melting Point ≈145°C (dec.)
Health Hazard 1 Warning
Synonym (S)-(-)-Carbamalactic acid; (S)-(-)-2-(Phenylaminocarbonyloxy)propionic acid
Chemical Name or Material (S)-(-)-2-(Phenylcarbamoyloxy)propionic acid
CAS 102936-05-0
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C10H11NO4
Assay 99%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.