missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-(+)-2-Phenylbutyric acid, 99%, Thermo Scientific Chemicals
$147.88 - $565.92
Chemical Identifiers
CAS | 4286-15-1 |
---|---|
Molecular Formula | C10H12O2 |
Molecular Weight (g/mol) | 164.204 |
MDL Number | MFCD00063164 |
InChI Key | OFJWFSNDPCAWDK-VIFPVBQESA-N |
Synonym | s-+-2-phenylbutyric acid, s-2-phenylbutanoic acid, 2s-2-phenylbutanoic acid, s-2-phenylbutyric acid, pubchem9874, 2-phenylbutyric acid, s, s--2-phenyl-mutyric acid, s-+-2-phenylbutanoic acid, s-+-2-phenyl-mutyric acid |
PubChem CID | 785329 |
IUPAC Name | (2S)-2-phenylbutanoic acid |
SMILES | CCC(C1=CC=CC=C1)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1275403
|
Thermo Scientific Chemicals
L1275403 |
1 g |
Each for $147.88
|
|
|||||
AAL1275406
|
Thermo Scientific Chemicals
L1275406 |
5 g |
Each for $565.92
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4286-15-1 | |
164.204 | |
OFJWFSNDPCAWDK-VIFPVBQESA-N | |
785329 | |
CCC(C1=CC=CC=C1)C(=O)O |
C10H12O2 | |
MFCD00063164 | |
s-+-2-phenylbutyric acid, s-2-phenylbutanoic acid, 2s-2-phenylbutanoic acid, s-2-phenylbutyric acid, pubchem9874, 2-phenylbutyric acid, s, s--2-phenyl-mutyric acid, s-+-2-phenylbutanoic acid, s-+-2-phenyl-mutyric acid | |
(2S)-2-phenylbutanoic acid |
Specifications
4286-15-1 | |
270°C to 272°C | |
C10H12O2 | |
MFCD00063164 | |
2045208 | |
OFJWFSNDPCAWDK-VIFPVBQESA-N | |
CCC(C1=CC=CC=C1)C(=O)O | |
164.204 | |
164.2 | |
(S)-(+)-2-Phenylbutyric acid |
1.055 | |
>110°C (230°F) | |
1.515 | |
1 g | |
s-+-2-phenylbutyric acid, s-2-phenylbutanoic acid, 2s-2-phenylbutanoic acid, s-2-phenylbutyric acid, pubchem9874, 2-phenylbutyric acid, s, s--2-phenyl-mutyric acid, s-+-2-phenylbutanoic acid, s-+-2-phenyl-mutyric acid | |
+100° (c=5 in Toluene) | |
(2S)-2-phenylbutanoic acid | |
785329 | |
99% |
Safety and Handling
GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only