missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-2-Octanol, 99%

Manufacturer:  Alfa Aesar™ L1242506

 View more versions of this product

Catalog No. AAL1242506



Description

Description

Applications
(S)-(+)-2-Octanol is used as chiral compounds.

Solubility
Soluble in water. (1.12 g/L) at 25°C

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents. Store at room temperature.
Specifications

Specifications

(S)-(+)-2-Octanol
Pleasant
179°C to 180°C
6192-12-7
MFCD00064283
14,6752
318.04
99%
76°C (169°F)
+10° (c=1 in Ethanol)
5g
C6H9O6Pr
1719323
KPXRPRLCONABEG-UHFFFAOYSA-K
130.23

Chemical Identifiers

6192-12-7
318.04
KPXRPRLCONABEG-UHFFFAOYSA-K
C6H9O6Pr
MFCD00064283
Safety and Handling

Safety and Handling

GHS H Statement
H319-H227
Causes serious eye irritation.
Combustible liquid.

GHS P Statement
P261-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

TSCA : TSCA

Documents