Please login to your online account to display your discounted pricing

(S)-(+)-2-Methylpiperazine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (S)-(+)-2-Methylpiperazine
CAS 74879-18-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H12N2
Formula Weight 100.16
MDL Number MFCD00171452
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC356260050

  SDS
thermo fisher scientific
356260050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder, Crystals and/or Chunks
Formula Weight 100.16
Quantity 5g
Color White to yellow
TSCA TSCA
Melting Point 86° to 91°C
Specific Rotation Condition + 13.50 (20.00°C c=1,toluene)
CAS Max % 100.0
Specific Rotation +5.8° to +7.8° (20°C, 589nm) (c=1, ethanol)
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Flammable solid.
CAS Min % 97.5
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material (S)-(+)-2-Methylpiperazine
CAS 74879-18-8
MDL Number MFCD00171452
Molecular Formula C5H12N2
Assay Percent Range 98%
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Flammable solid.