Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-2-Methylpiperazine, 98+%

Chemical Identifiers

Chemical Name or Material (S)-(+)-2-Methylpiperazine
CAS 74879-18-8
Molecular Formula C5H12N2
Formula Weight 100.17
MDL Number MFCD00171452
UN Number UN2925
Beilstein 3647995
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1442903

 
alfa aesar™
L1442903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL14429MD

 
alfa aesar™
L14429MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 100.17
Chemical Name or Material (S)-(+)-2-Methylpiperazine
Assay ≥98%
MDL Number MFCD00171452
CAS 74879-18-8
Beilstein 3647995
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Health Hazard 3 GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Boiling Point 155° to 156°C
Quantity 1g
Flash Point 65°C (149°F)
Molecular Formula C5H12N2
UN Number UN2925
Melting Point 89° to 91°C
Optical Rotation +7° (c=1 in Ethanol)
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.