Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-2-Methyl-CBS-oxazaborolidine monohydrate, 94%

$192.41 - $608.01

Chemical Identifiers

Chemical Name or Material (S)-2-Methyl-CBS-oxazaborolidine monohydrate
CAS 112022-81-8
Molecular Formula C18H20BNO·H2O
Formula Weight 295.19 (277.17 Anhydrous)
MDL Number MFCD00078439
Beilstein 8492123
Synonym (S)-3,3-Diphenyl-1-methylpyrrolidino[1,2-c]-1,3,2-oxazaborole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0921903

 
alfa aesar™
L0921903
1g Each for $608.01

AAL09219MD

 
alfa aesar™
L09219MD
250mg Each for $192.41
Description & Specifications

Specifications

Formula Weight 295.19 (277.17 Anhydrous)
Chemical Name or Material (S)-2-Methyl-CBS-oxazaborolidine monohydrate
Assay 94%
Molecular Formula C18H20BNO·H2O
CAS 112022-81-8
MDL Number MFCD00078439
Beilstein 8492123
Synonym (S)-3,3-Diphenyl-1-methylpyrrolidino[1,2-c]-1,3,2-oxazaborole
Melting Point 115° to 121°C
Quantity 1g
Optical Rotation −67° (c=1 in Methanol)