Please login to your online account to display your discounted pricing

(S)-(-)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene, ACROS Organics™


Quantity 5mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC369770050

 View more versions of this product

Catalog No. AC369770050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 277.17
Quantity 5mL
Density 0.9290g/mL
Chemical Name or Material (S)-(-)-2-Methyl-CBS-oxazaborolidine
Name Note 1M solution in toluene
Appearance of Solution Colorless to amber solution
Molecular Formula C18H20BNO
MDL Number MFCD00078439
CAS 112022-81-8
CAS Min % 28.34
CAS Max % 32.82
CAS 108-88-3
CAS Min % 67.18
CAS Max % 71.66
Packaging Glass bottle
Synonym (S)-3,3-Diphenyl-1-methylpyrrolidino[1,2-c]-1,3,2-oxazaborole
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye damage.
Suspected of damaging the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizzi
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (S)-(-)-2-Methyl-CBS-oxazaborolidine
Name Note 1M solution in toluene
CAS 112022-81-8
CAS Min % 28.34
CAS Max % 32.82
CAS 108-88-3
CAS Min % 67.18
CAS Max % 71.66
Molecular Formula C18H20BNO
Formula Weight 277.17
MDL Number MFCD00078439
Synonym (S)-3,3-Diphenyl-1-methylpyrrolidino[1,2-c]-1,3,2-oxazaborole
Accessories

Danger

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye damage.
Suspected of damaging the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizzi

SDS & Safety Documents