missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-+-2-(Methoxymethyl)pyrrolidine, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC309910025

 View more versions of this product

Catalog No. AC309910025

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
115.18
Clear colorless to yellow
0.9300g/mL
+35.6° to +37.6° (20°C, 589nm) (c=2, 2N HCl)
98.5
1.4450 to 1.4470 (20°C, 589nm)
(S)-(+)-2-(Methoxymethyl)pyrrolidine
63126-47-6
99%
Liquid
2.5g
+ 36.60 (20.00°C c=2,2N HCl)
100.0
Authentic
Glass bottle
o-Methyl-L-prolinol
MFCD00010408
C6H13NO
99%

Chemical Identifiers

63126-47-6
100.0
C6H13NO
MFCD00010408
(S)-(+)-2-(Methoxymethyl)pyrrolidine, 99%
98.5
99%
115.18
o-Methyl-L-prolinol
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA