Please login to your online account to display your discounted pricing

(S)-+-2-(Methoxymethyl)pyrrolidine, 99%, ACROS Organics™

$87.61 - $87.61

Chemical Identifiers

Chemical Name or Material (S)-(+)-2-(Methoxymethyl)pyrrolidine
Name Note 99%
CAS 63126-47-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H13NO
Formula Weight 115.18
MDL Number MFCD00010408
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC309910025

  SDS
thermo fisher scientific
AC309910025
Glass bottle 2.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC309915000

  SDS
thermo fisher scientific
309915000
Glass bottle 500mg Each for $87.61
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 115.18
Quantity 2.5g
Color Clear colorless to yellow
TSCA TSCA
Specific Rotation Condition + 36.60 (20.00°C c=2,2N HCl)
Density 0.9300g/mL
CAS Max % 100.0
Specific Rotation +35.6° to +37.6° (20°C, 589nm) (c=2, 2N HCl)
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
Packaging Glass bottle
Refractive Index 1.4450 to 1.4470 (20°C, 589nm)
Synonym o-Methyl-L-prolinol
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Chemical Name or Material (S)-(+)-2-(Methoxymethyl)pyrrolidine
MDL Number MFCD00010408
CAS 63126-47-6
Molecular Formula C6H13NO
Name Note 99%
Assay Percent Range 99%
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.