Please login to your online account to display your discounted pricing

(S)-+-2-(Methoxymethyl)pyrrolidine, 99%, ACROS Organics™


Quantity 500mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  309915000

 View more versions of this product

Catalog No. AC309915000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 115.18
Physical Form Liquid
Quantity 500mg
Color Clear colorless to yellow
TSCA TSCA
Density 0.9300g/mL
Specific Rotation Condition + 36.60 (20.00°C c=2,2N HCl)
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
CAS Min % 98.5
Specific Rotation +35.6° to +37.6° (20°C, 589nm) (c=2, 2N HCl)
Refractive Index 1.4450 to 1.4470 (20°C, 589nm)
Synonym o-Methyl-L-prolinol
Name Note 99%
MDL Number MFCD00010408
Molecular Formula C6H13NO
Assay Percent Range 99%
CAS 63126-47-6
Chemical Name or Material (S)-(+)-2-(Methoxymethyl)pyrrolidine

Specifications

Chemical Name or Material (S)-(+)-2-(Methoxymethyl)pyrrolidine
Name Note 99%
CAS 63126-47-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H13NO
Formula Weight 115.18
MDL Number MFCD00010408
Synonym o-Methyl-L-prolinol
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

SDS & Safety Documents