Please login to your online account to display your discounted pricing

(S)-2-Isopropylsuccinic acid-1-methyl ester, 95%, 98% ee, ACROS Organics™

$1,247.10 - $1,247.10

Chemical Identifiers

Chemical Name or Material (S)-2-Isopropylsuccinic acid-1-methyl ester
Name Note 95%, (98% e.e.)
CAS 208113-95-5
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C8H14O4
Formula Weight 174.19
MDL Number MFCD01091031
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC362110010

  SDS
fisher scientific acros
362110010
Glass bottle 1g Each for $1,247.10
Description & Specifications

Specifications

Assay ≥94%
Physical Form Viscous Liquid
Formula Weight 174.19
Quantity 1g
Color Clear yellow
TSCA TSCA
CAS Max % 100.0
CAS Min % 94.0
Synonym (S)-3-Methoxycarbonyl-4-methylpentanoic acid
Infrared Spectrum Authentic
Packaging Glass bottle
Enantiomeric Excess 97.5% min. (GC)
Name Note 95%, (98% e.e.)
Molecular Formula C8H14O4
MDL Number MFCD01091031
CAS 208113-95-5
Chemical Name or Material (S)-2-Isopropylsuccinic acid-1-methyl ester
Assay Percent Range 95%