Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-2-Isobutylsuccinic acid 1-methyl ester, 98+%, ee 98+%

$147.22 - $147.22

Chemical Identifiers

Chemical Name or Material (S)-(-)-2-Isobutylsuccinic acid 1-methyl ester
CAS 213270-36-1
Molecular Formula C9H16O4
Formula Weight 188.22
MDL Number MFCD01091033
Beilstein 8110824
Synonym (S)-3-Methoxycarbonyl-5-methylhexanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL18268MD

  SDS
alfa aesar™
L18268MD
250mg Each for $147.22
Description & Specifications

Specifications

Formula Weight 188.22
CAS 213270-36-1
Molecular Formula C9H16O4
Chemical Name or Material (S)-(-)-2-Isobutylsuccinic acid 1-methyl ester
Synonym (S)-3-Methoxycarbonyl-5-methylhexanoic acid
Quantity 250mg
Beilstein 8110824
Assay Percent Notes ee ≥98%
MDL Number MFCD01091033
Assay ≥98%

Applications
(S)-(-)-2-Isobutylsuccinic acid 1-methyl ester is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.