missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-2-Hydroxy-4-phenylbutyric Acid, TCI America™

$146.67 - $508.77

Chemical Identifiers

CAS 115016-95-0
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD06656492
InChI Key JNJCEALGCZSIGB-VIFPVBQESA-N
PubChem CID 11159656
IUPAC Name (2S)-2-hydroxy-4-phenylbutanoic acid
SMILES C1=CC=C(C=C1)CCC(C(=O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H13391G View Documents TCI America
H13391G
1g Each for $146.67
 
H13395G View Documents TCI America
H13395G
5g Each for $508.77
 
Specifications

Chemical Identifiers

115016-95-0
180.203
JNJCEALGCZSIGB-VIFPVBQESA-N
(2S)-2-hydroxy-4-phenylbutanoic acid
C10H12O3
MFCD06656492
11159656
C1=CC=C(C=C1)CCC(C(=O)O)O

Specifications

White
1g
C10H12O3
JNJCEALGCZSIGB-VIFPVBQESA-N
(2S)-2-hydroxy-4-phenylbutanoic acid
11159656
Crystal-Powder at 20°C
(S)-2-Hydroxy-4-phenylbutyric Acid
117°C
115016-95-0
MFCD06656492
C1=CC=C(C=C1)CCC(C(=O)O)O
180.203
180.20
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-4133

TSCA : No