missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(+)-2-Hydroxy-3-methylbutanoic acid, 99+%, Thermo Scientific™

Chemical Identifiers

CAS 17407-55-5
Molecular Formula C5H9O3
Molecular Weight (g/mol) 117.13
MDL Number MFCD00066443
InChI Key NGEWQZIDQIYUNV-BYPYZUCNSA-M
Synonym s-+-2-hydroxy-3-methylbutyric acid, s-2-hydroxy-3-methylbutanoic acid, l-alpha-hydroxyisovaleric acid, s-2-hydroxy-3-methylbutyric acid, 2s-2-hydroxy-3-methylbutanoic acid, unii-rub6h7st9k, s-2-hydroxyisovaleric acid, s-+-2-hydroxy-3-methylbutanoic acid, butanoic acid, 2-hydroxy-3-methyl-, 2s, rub6h7st9k
PubChem CID 853180
ChEBI CHEBI:60631
IUPAC Name (2S)-2-hydroxy-3-methylbutanoic acid
SMILES CC(C)[C@H](O)C([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC269900010
SDS View Documents Promotion Details
Thermo Scientific Chemicals
269900010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC269900050
SDS View Documents Promotion Details
Thermo Scientific Chemicals
269900050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

17407-55-5
117.13
NGEWQZIDQIYUNV-BYPYZUCNSA-M
853180
(2S)-2-hydroxy-3-methylbutanoic acid
C5H9O3
MFCD00066443
s-+-2-hydroxy-3-methylbutyric acid, s-2-hydroxy-3-methylbutanoic acid, l-alpha-hydroxyisovaleric acid, s-2-hydroxy-3-methylbutyric acid, 2s-2-hydroxy-3-methylbutanoic acid, unii-rub6h7st9k, s-2-hydroxyisovaleric acid, s-+-2-hydroxy-3-methylbutanoic acid, butanoic acid, 2-hydroxy-3-methyl-, 2s, rub6h7st9k
CHEBI:60631
CC(C)[C@H](O)C([O-])=O

Specifications

17407-55-5
100.0
99% min. (GC)
C5H9O3
MFCD00066443
+18° (20°C c=1,CHCl3)
s-+-2-hydroxy-3-methylbutyric acid, s-2-hydroxy-3-methylbutanoic acid, l-alpha-hydroxyisovaleric acid, s-2-hydroxy-3-methylbutyric acid, 2s-2-hydroxy-3-methylbutanoic acid, unii-rub6h7st9k, s-2-hydroxyisovaleric acid, s-+-2-hydroxy-3-methylbutanoic acid, butanoic acid, 2-hydroxy-3-methyl-, 2s, rub6h7st9k
CC(C)[C@H](O)C([O-])=O
117.13
CHEBI:60631
99+%
99.0
Authentic
Glass bottle
CH3CH(CH3)CH(OH)COOH
1g
+ 18.00
NGEWQZIDQIYUNV-BYPYZUCNSA-M
(2S)-2-hydroxy-3-methylbutanoic acid
853180
118.13
(S)-(+)-2-Hydroxy-3-methylbutanoic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only